Halyard QUICK CHECK sterilization wraps are the fast, easy way to ensure instrument sterility is uncompromised.
- Two distinct, contrasting layers, translucent and opaque are bonded together making inspecting trays easier and faster than ever
- Superior microbial barrier protection with Powerguard* technology
- Cleared for all of the same sterilization systems as Halyard ONE-STEP Wrap
- Made from recyclable #5 polypropylene fabric
To make specific trays stand out from the rest, simply flip the sheet and wrap with the other side out. Same protection, no protocol changes. QUICK CHECK Wraps give the same time savings of Halyard ONE-STEP Wraps, along with protection and performance you expect. Like all Halyard Wraps, QUICK CHECK provide superior microbial barrier protection with POWERGUARD technology, plus it’s cleared for all of the same sterilization systems as Halyard ONE-STEP Wrap. Because it’s ONE-STEP, it’s easy to make the switch to QUICK CHECK Wrap. So, you can enjoy the benefits of this innovative wrap without having to retrain staff or change protocols.