Would you like to log in again?
Continuous Liner Kits, 14" Diameter, Rheo Flexibles (Division of Rheo Engineering)
Supplier: Rheo Flexibles
4000683 4000523 4000520 4001012
76559-106EA 0 USD InStock
76559-106 76557-586 76559-114 76557-778 76559-112 76557-676 76557-698 76559-110
Continuous Liner Kits, 14" Diameter, Rheo Flexibles (Division of Rheo Engineering)
Mailers and Shippers
The Rheo Continuous Liner is an excellent pack-off solution for transferring materials from a work process into a transport container while maintaining a high level of containment. Continuous Liners come in various sizes and can be configured to work with existing process equipment.
  • Constructed from RheoFlex 302 polyethylene material
  • FDA approved for packaging
  • USP and EP compliant
  • 21 CFR 177.1520 compliant
  • Matte finish
  • Anti-static film
Continuous Liners keep the powder pack-off process clean and reduce the risk of cross contamination within the process suite. This technology can also provide a safe bag-in/bag-out option for transferring items into and out of an isolator.
Certifications: USP <661.1>, USP <661.2>, USP <88>, USP <87>, USP Class VI, EP 3.1.3, EU 10/2011, 94/62/EC, CONEG.
Ordering information: Store in box, out of UV light, in a cool and dry location.
Delivery information: Continuous Liner Kit comes with a Continuous Liner Pack with Support Ring, Stainless Steel Canister, Tri-Clamp with Support Flanges, and GripLoc Clamp. The Continuous Liner Pack is the replacement liner.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty