Would you like to log in again?
Ceramic Magnetic Hotplate Stirrer, HSC, VELP Scientifica
Supplier: VELP Scientifica
F20510552
76559-352EA 613.07 USD InStock
76559-352
Ceramic Magnetic Hotplate Stirrer, HSC, VELP Scientifica
Stirrers Magnetic Hotplate Stirrers
The entry level hotplate stirrer for general laboratory applications. You can rely on the HSC for everyday laboratory applications like sample and buffer solutions preparation, as it stirs volumes up to 15 liters (H₂O). HSC is equipped with a cutting-edge Heating Plate technology engineered by VELP that ensures temperature homogeneity, thermal efficiency and durable heating performance over time.
  • Analog hotplate stirrer with ceramic plate and techno polymer housing
  • Features ensure a high level of safety and protection for researchers and laboratory operators
  • HSC stirs aqueous solutions from 50 up to 1300 rpm, an ideal solution for general laboratory applications
  • The extremely resistant ceramic plate is easy to clean and resistant to acids, bases and solvents
  • Monophase PCM motor
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty