Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 
NEBNext FFPE DNA Repair v2 Module, New England Biolabs
Supplier: New England Biolabs
NEBNext®
E7360S E7360L
76559-400EA 616 USD InStock
76559-400 76559-398
NEBNext FFPE DNA Repair v2 Module, New England Biolabs
Nucleic Acid Reagents Next Generation Sequencing Reagents
This new product further improves repair of FFPE DNA in next generation sequencing (NGS) workflows. The methods used for fixation and storage of Formalin-Fixed, Paraffin-Embedded (FFPE) DNA samples cause significant damage, making it challenging to obtain high quality sequence data.
  • Higher repair efficiency with FFPE DNA
  • A more streamlined workflow for NGS library preparation
  • Convenient reaction buffer for efficient FFPE DNA repair, end repair and dA-tailing
The NEBNext FFPE DNA Repair v2 Module is an optimized cocktail of enzymes designed to repair FFPE DNA, and supplied with optimized reagents to enable a streamlined workflow for NGS library preparation.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty