Would you like to log in again?
Kimtech™ G3 Sapphire Nitrile Cleanroom Gloves
Supplier: Kimberly-Clark
Kimtech™
55875 55876 55878 55877
76575-512CS 338.9 USD InStock
76575-512 76575-514 76575-516 76575-508
Kimtech™ G3 Sapphire Nitrile Cleanroom Gloves
Gloves
Sapphire blue nitrile cleanroom gloves protect against contamination for products and processes in cleanroom manufacturing.
  • Powder-free
  • Straight fingers
  • Ambidestrious
  • Textured fingertip
  • Beaded cuff
  • Double chlorinated
  • Laundered
  • Bulk packaged
Gloves are designed for cleanroom/industrial use only. They meet FDA food contact requirements set forth in 21 CFR 177.2600.
Packaging: Gloves are packaged 100 per pack, 1000 gloves per case.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty