Would you like to log in again?
Human Cytokeratin 18-M65 (CK 18-M65) ELISA Kit, AFG Bioscience
Supplier: AFG Bioscience
EK700484
76576-976EA 426.89 USD InStock
76576-976
Human Cytokeratin 18-M65 (CK 18-M65) ELISA Kit, AFG Bioscience
Assays ELISAs
  • Assay principle: Quantitative
  • Research topic: Signal transduction
  • Abbreviation: CK 18-M65
  • Detection wavelength: 450 nm
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty