Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Bovine Tumor Necrosis Factor Receptor Superfamily Member 1A (TNFRSF1A) ELISA Kit, AFG Bioscience

Supplier: AFG Bioscience
EK760256
76577-746EA 426.89 USD InStock
76577-746
Bovine Tumor Necrosis Factor Receptor Superfamily Member 1A (TNFRSF1A) ELISA Kit, AFG Bioscience
Assays ELISAs
  • Research Topic: Cancer
  • Abbreviation: TNFRSF1A
  • Detection Wavelength: 450 nm

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty