Would you like to log in again?
Bovine borrelia burgdorferi (Bb) Antibody (IgM) ELISA Kit, AFG Bioscience
Supplier: AFG Bioscience
EK760304
76577-904EA 426.89 USD InStock
76577-904
Bovine borrelia burgdorferi (Bb) Antibody (IgM) ELISA Kit, AFG Bioscience
Assays ELISAs
  • Assay Principle: Qualitative
  • Assay Type: Capture
  • Abbreviation: Bb
  • Detection Wavelength: 450 nm
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty