Would you like to log in again?
Human anti-Complement Factor H (anti-CFH) ELISA Kit, AFG Bioscience
Supplier: AFG Bioscience
EK711242
76578-240EA 426.89 USD InStock
76578-240
Human anti-Complement Factor H (anti-CFH) ELISA Kit, AFG Bioscience
Assays ELISAs
  • Assay principle: Quantitative
  • Research topic: Immunology
  • Abbreviation: anti-CFH
  • Detection wavelength: 450 nm
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty