Would you like to log in again?
Illuminated Tissue Flotation Bath and Slide Drying Work Station Bundle, Mopec
Supplier: Mopec
BK250
76579-220EA 4262.77 USD InStock
76579-220
Illuminated Tissue Flotation Bath and Slide Drying Work Station Bundle, Mopec
Slide Warmers/Dryers
Mopec’s illuminated tissue flotation bath features an LED light and high-contrast background that allows users to better view tissue sections while also detecting foreign particles floating on the surface of the water.
  • Designed to take up minimal bench-top space
  • Slide drying mini workstation fits directly over illuminated tissue flotation bath
  • Digital temperature control
  • Rapid heating system ensures system is ready to use in less time
  • Anti-microbial coating to help prevent bacteria and virus growth
This workstation features a mini hotplate to remove wrinkles from tissue sections as well as a sloped drying area that holds up to 12 slides.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty