Would you like to log in again?
Slide Drying Hotplate, Mopec
Supplier: Mopec
BK400 BK400-L
76579-276EA 2089.04 USD InStock
76579-276 76579-278
Slide Drying Hotplate, Mopec
Slide Warmers/Dryers
Mopec’s slide drying hotplate is designed to quickly and efficiently dry paraffin tissue sections. The rapid heating system ensures the hotplate is ready to use in less time, while the digital temperature control allows for accurate control of temperatures.
  • Designed to take up minimal bench-top space
  • Digital temperature control to meet precise temperature requirements
  • Rapid heating system ensures system is ready to use in less time
  • Slide support rails can be adjusted to fit different slide sizes
  • Anti-microbial coating to help prevent bacteria and virus growth
  • Optional lid helps reduce drying time and improve temperature control (BK400-L)
Slides are places on the four moveable rails at an angle to allow water drain off, while heated air passes underneath the slides to reduce drying times. The system features an optional lid to reduce drying times and improve temperature control. Made from stainless steel and can be used while slides are on the hotplate.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty