Would you like to log in again?
Large Slide Drying Hotplate, Mopec
Supplier: Mopec
BK401-L BK401
76579-280EA 2651.81 USD InStock
76579-280 76579-282
Large Slide Drying Hotplate, Mopec
Slide Warmers/Dryers
Mopec’s large slide drying hotplate is designed to quickly and efficiently dry paraffin tissue sections. The rapid heating system ensures the hotplate is ready to use in less time, while the digital temperature control allows for accurate control of temperatures.
  • Designed to take up minimal bench-top space
  • Digital temperature control to meet precise temperature requirements
  • Rapid heating system ensures system is ready to use in less time
  • Slide support rails can be adjusted to fit different slide sizes
  • Anti-microbial coating to help prevent bacteria and virus growth
  • Optional lid helps reduce drying time and improve temperature control (BK400-L)
Slides are placed on the six moveable rails at an angle to allow water drain off, while heated air passes underneath the slides to reduce drying times. The system features an optional lid to reduce drying times and improve temperature control. Made from stainless steel and can be used while slides are on the hotplate.
Caution: The product must be placed on a smooth, level, and sturdy work surface. Suitable for use in ambient temperatures from 5 to 40 °C with a maximum humidity 80% (temperature 31 °C) decreasing to 50% (temperature 40 °C).
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty