Would you like to log in again?
Paraffin Wax Dispensers, Mopec
Supplier: Mopec
BK575X2 BK575
76579-286EA 2841.97 USD InStock
76579-286 76579-288
Paraffin Wax Dispensers, Mopec
Paraffin Wax Trimmers and Dispensers
The Mopec paraffin wax dispenser is designed to take up minimal bench-top space while providing a rapid heating system that produces melted paraffin wax in only 30 minutes.
  • Designed to take up minimal bench-top space
  • Digital temperature control
  • 15 L has two separate tanks with independent heating controls
  • Rapid heating system ensures system is ready to use in less time
  • Heated tap prevents wax buildup
  • Non-drip lever with three positions
  • Double-layered tanks provide internal temperature control and ensures outer surfaces are safe to touch
  • Anti-microbial coating to help prevent bacteria and virus growth
The dispenser features a non-drip lever, a dedicated heating system for the tap to prevent wax blockages, and is available in 7.5 L and 15 L capacities. Units sold separately.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty