Would you like to log in again?
High Capacity Section Dryer, Mopec
Supplier: Mopec
BK701-T2 BK701-T1 BK701-R BK701
76579-304EA 5287.78 USD InStock
76579-304 76579-306 76579-308 76579-310
High Capacity Section Dryer, Mopec
Slide Warmers/Dryers
Mopec’s high capacity section dryer is the perfect solution for high workload laboratories. Similar to the mini section dryer, it’s designed to efficiently and rapidly blow temperature-controlled air through the base of the instrument to dry specimens without the risk of overheating.
  • Fan-assisted heating dramatically reduces drying time
  • Designed to take up minimal bench-top space
  • Digital temperature control
  • Rapid heating system ensures system is ready to use in less time
  • Anti-microbial coating to help prevent bacteria and virus growth
  • Rack holder (BK701-R) holds 8 slide racks and works in conjunction with timer units
  • Timer units (BK701-T1 and BK701-T2) allow monitoring of four slide racks each and ensures that each rack has the correct amount of time in the dryer
Featuring a large chamber, the system can house up to 30 slides within each of its 13 slide racks. The transparent lid can be positioned at any angle, allowing users to use both hands to place or remove items from the dryer. Additional options include a rack holder and two timer units.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty