Synonyms:
Methylaldehyde
, Formol
, Paraformaldehyde aqueous solution
, Formaldehyde
BG507
BG509
BG500
BG512
BG502
BG518
BG548
BG504
BG515
BG505
BG524
BG506
BG516
76579-330EA
56.04
USD
InStock
76579-330
76579-320
76579-332
76579-344
76579-322
76579-312
76579-334
76579-324
76579-326
76579-316
76579-338
76579-328
76579-318
Formalin prefilled 10%
Formalin
Mopec’s Prefilled Formalin Containers are specifically designed to safely collect, store and transport various sizes of tissue specimens. Containers are half-filled with a 10% neutral buffered formalin fixative and come in a variety of sizes.
- Filled with a 10% neutral buffered formalin
- General purpose fixative for tissues.
- Penetrates quickly, but fixes slowly
- No other dilution or additional agents are necessary