Would you like to log in again?
Formaway Green Liquid Formalin Neutralizer, Mopec
Supplier: Mopec
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
BG560-5 BG560-1 BG560-2.5
76579-380EA 114.81 USD InStock
76579-380 76579-382 76579-378
Formaway Green Liquid Formalin Neutralizer, Mopec
Stains Microscopy and staining reagents
Mopec’s Formaway green liquid formalin neutralizer quickly and effectively neutralizes spent formalin for sewer disposal. Simply add 32 ounces of Formaway green to each gallon of 10% neutral buffered formalin waste and allow to stand for at least 4 hours before disposing.
  • Sold individually
Collect formalin waste in the Formaway collection container or equivalent container. Use a funnel with a 'screen insert' while collecting waste to remove tissue debris from waste formalin. Add 32-fl oz. of Formaway green to each gallon (128-fl oz.) of 10% NBF formalin waste. (For metric users, add 1 liter of Formaway green to 4 liters of 10% NBF formalin waste.) Secure lid on waste collection container and agitate container to mix solutions. Allow mixed solutions to stand for approximately 4 hours, adequately reducing the formalin in the waste solution to a non hazardous safe disposal into the sanitary sewer. Repeat step #4, decreasing any potential exposure to the headspace vapor in the collection container. Test treated solution with rapid QC analysis and carefully pour treated waste into the sanitary sewer. Rinse container with cold tap water, preparing it for re-use.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty