Would you like to log in again?
Biohazard Specimen Transport Bags, Mopec
Supplier: Mopec
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
BG684 BG682 BG682-P BG683
76579-402CS 219.92 USD InStock
76579-402 76579-404 76579-398 76579-400
Biohazard Specimen Transport Bags, Mopec
Bags Sample Bags
Mopec’s biohazard specimen transport bags are the perfect solution for safely and securely transporting various blood tubes, urine containers, or pathology specimens.
  • Used for transporting blood tubes, urine containers, or pathology specimens.
  • Ideal for use in hospitals and laboratories
  • Made from a high-quality LDPE material
  • Re-closable zipper
  • Separate pouch to safely store documents
  • Feature 'Biohazard' symbol
Ideal for use in hospitals and laboratories, these bags are made from a high-quality LDPE material, feature a re-closable zipper and have a separate pouch to safely store documents. Available in three sizes. 100 bags/pack. Sold by the case of 10 packs.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty