Would you like to log in again?
VWR® Autoclavable Cleanroom Goggles
Supplier: VWR
VWR®
G14ACAFL G14ACAF
76579-452CS 1183.45 USD InStock
76579-452 76579-454
VWR® Autoclavable Cleanroom Goggles
Goggles
Cleanroom goggles feature a white frame with indirect ventilation.
  • Anti-fog and anti-scratch protection
  • Clear lens
  • Indirect ventilation
  • Silicone strap
  • White frame
  • Autoclavable
  • Tested to withstand >40 autoclave cycles (30 minutes duration at 121/250 °C)
Clear replacement lenses also feature anti-fog and anti-scratch protection.
Ordering information: Replacement lenses are sold separately.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty