Would you like to log in again?
Super-Safe™ Carton Opener, Slice®
Supplier: Slice
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
10477
76579-608BX 274.77 USD InStock
76579-608
Super-Safe™ Carton Opener, Slice®
Knives
Super-Safe™ carton opener is a unique knife with a design that is ideal for those with dexterity issues or anyone wearing work gloves. Its large, circular handle fits comfortably in the palm for straight, flush cuts. It features a finger-friendly® blade that is safe to the touch, but cuts effectively.
  • Automatic spring-loaded retraction for added safety
  • Ambidextrous design
  • Non-sparking, non-conductive, non-magnetic blade
  • Chemically inert blade never rusts
  • Oil and lubricant free blade
  • Blade edge lasts up to 11× longer than metal
  • No-tool blade change
The Slice® Super-Safe™ carton opener is a unique knife with a design that is ideal for those with dexterity issues or anyone wearing work gloves. Its large, circular handle fits comfortably in the palm for straight, flush cuts. Press the orange button with an index finger to unlock the blade guard. Then, press the tool down, flush, onto the cutting surface and pull to make the cut. Once the 10477 is no longer held against the cutting surface, its blade guard redeploys, keeping the user safe in the event of a slip. As with all Slice tools, the Super-Safe™ carton opener features a finger-friendly® blade that is safe to the touch, cuts effectively, and lasts up to 11 times longer than comparable steel blades. The installed rounded-tip blade (10526) can also be replaced with an optional pointed-tip blade (10528) for materials that require puncturing to initiate the cut.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty