Would you like to log in again?
Mini Cleaver, Slice®
Supplier: Slice
10497
76579-616BX 274.91 USD InStock
76579-616
Mini Cleaver, Slice®
Knives
Slice designed a thicker handle with soft touch overlay for a comfortable, sturdy grip. Mini cleaver has features like a lanyard hole and blade guard storage. The advanced-ceramic blade features Slice’s proprietary finger-friendly® edge, which is safe to the touch and lasts up to 11 times longer than steel blades.
  • Ambidextrous design
  • Finger-friendly® blade cuts effectively
  • Non-conductive, non-magnetic blade
  • Chemically inert blade never rusts
  • Oil and lubricant free blade
  • 100% zirconium oxide blade
  • Fewer blade changes - fewer injuries
This mini cleaver brings Slice® quality to a different kind of handheld cutting tool. Whether you are cutting thin wires, lab samples, or manufacturing components, you will appreciate its hand feel. Slice designed a thicker handle with soft touch overlay for a comfortable, sturdy grip. The Mini Cleaver has features like a lanyard hole and blade guard storage in the handle, so it is as convenient as it is robust. The 10497 comes with a rounded-tip utility blade and is compatible with our pointed-tip and serrated utility blades (10524, 10525, 10523 respectively). The advanced-ceramic blade features Slice’s proprietary finger-friendly® edge, which is safe to the touch and lasts up to 11 times longer than comparable steel blades. The chemically inert blade can be autoclaved and never rusts.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty