Would you like to log in again?
OsmoTECH® XT Single-Sample Micro-Osmometer, Advanced Instruments
Supplier: Advanced Instruments
OsmoTECH® XT
OSMOTECH_XT
76579-780EA 81261.4 USD InStock
76579-780
OsmoTECH® XT Single-Sample Micro-Osmometer, Advanced Instruments
Clinical Diagnostic Supplies Clinical Analysers
Increase your productivity and reliability with powerful freezing point depression technology for osmolality measurement with the OsmoTECH® XT single-sample osmometer.
  • Offers the widest range of osmolality testing (0 to 4000 mOsm/kg H₂O)
  • Large user-friendly interface​
  • New, intelligent freezing technology
  • Fully compliant with 21 CFR part 11, GMP, and EU Annex 11 with e-Signature
  • Meets Pharmacopeia osmolality testing guidelines
The OsmoTECH® XT measures the widest variety of sample types (high concentration, high viscosity) accurately across bioprocessing and cell and gene therapy workflows. It’s also designed to make your work as streamlined and comfortable as possible with even more robust data integrity and compliance features to match your digital environment. See how the OsmoTECH® XT osmometer will change the way you work in the lab.
Accessories information: OsmoTECH® XT Single-Sample Micro-Osmometer IQ/OQ Service (sold separately): Our experts developed this comprehensive qualification package to comply with USP <1058> Analytical Instrument Qualification and USP <785> Osmolality and Osmolarity Testing. Don’t go it alone. Regulated environments require documented proof that your manufacturing process consistently produces quality product. Performing qualification internally is costly, time-consuming, and problematic. Our expert qualification services ensure fast and reliable commissioning of your instrument. Eliminate delays with our IQ/OQ. Our comprehensive services ensure no workflow interruption so you can focus on your work.
OsmoTECH® XT Single-Sample Micro-Osmometer Installation and Training (sold separately): Our installation and training experts ensure that your instrument is properly installed and your staff is properly trained. Training topics include installation, operation, and maintenance. Benefits of installation and training include: proper installation and setup, unpacking and inspection, Instrument training for staff, Inspector-ready training documentation. Our installation and training experts ensure that your instrument is properly installed and your staff is properly trained.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty