Would you like to log in again?
Cole-Parmer® Essentials Color-Coded Cable/Zip Ties
Supplier: Cole Parmer
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
06842-32 06842-40 06842-31 06842-39 06842-36 06842-38 06842-33 06842-41 06830-63 06830-41 06842-30 06830-61 06830-31 06830-62 06830-32 06830-33 06842-35 06842-37 06842-34 06842-29 06830-64 06830-65
76579-872CS 1.8 USD InStock
76579-872 76579-896 76579-940 76579-874 76579-820 76579-974 76579-930 76579-810 76579-844 76579-976 76579-900 76579-966 76579-978 76579-868 76579-934 76579-946 76579-816 76579-904 76579-828 76579-938 76579-906 76579-818
Cole-Parmer® Essentials Color-Coded Cable/Zip Ties
Ties
These ties are injection-molded of ⁶/₆ nylon, and won’t corrode or stretch.
  • Ties hold securely, even in vibrating, oily, or wet environments
  • Easily apply ties by hand or use a tensioning tool
Ties are UL-component recognized, and are self-extinguishing as per UL 94 Flam. Class: V-2.
Ties withstand temperatures to 85 °C (185 °F ) and have a melting point of 257 °C (495 °F). All ties remain flexible to −40 °C (−104 °F). Efficient range of ties may be higher or lower depending upon specific applications.
Ordering information: Tools must be ordered separately.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty