Cable ties won’t corrode or stretch.
- Holds securely, even in vibrating, oily, or wet environments
- Withstand temperatures to 85 °C (185 °F)
These ties are injection-molded of 6/6 nylon, and won’t corrode or stretch. Durable locking mechanism holds securely even when used to bundle vibrating or flexing materials. Ties continue to hold tension even in oily spray or wet environments. Easily apply ties by hand or use a tensioning tool (tools must be ordered separately).
Ties are UL-component recognized, and are self-extinguishing as per UL 94 flam. Class: V-2.
Ties withstand temperatures to 85 °C (185 °F) and have a melting point of 257 °C (495 °F). All ties remain flexible to −40 °C (−104 °F). Efficient range of ties may be higher or lower depending upon specific applications.