Would you like to log in again?
Cole-Parmer® Essentials Cable or Zip Ties, Black and White
Supplier: Cole Parmer
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
06842-28 06830-48 06830-49 06842-25 06830-43 06842-19 06830-58 06842-05 06830-66 06842-12 06842-14 06830-36 06842-15 06830-52 06830-42 06842-21 06830-39 06842-22 06830-46 06842-11 06830-88 06842-06 06842-24 06842-08 06830-29 06842-26 06842-01 06842-23 06842-10 06842-07 06842-03 06842-02 06830-25 06842-04 06842-18 06830-20 06842-13 06830-68 06842-17 06842-00 06830-78 06842-20 06830-91 06842-27 06842-16
76579-880BG 16.86 USD InStock
76579-880 76579-980 76579-860 76579-882 76579-982 76579-840 76579-884 76579-852 76579-952 76579-854 76579-832 76579-876 76579-898 76579-932 76579-910 76579-954 76579-812 76579-834 76579-856 76579-814 76579-836 76579-858 76579-936 76579-918 76579-870 76579-970 76579-850 76579-972 76579-950 76579-984 76579-864 76579-886 76579-842 76579-942 76579-920 76579-888 76579-822 76579-944 76579-824 76579-924 76579-826 76579-848 76579-948 76579-928 76579-908
Cole-Parmer® Essentials Cable or Zip Ties, Black and White
Ties
Cable ties won’t corrode or stretch.
  • Holds securely, even in vibrating, oily, or wet environments
  • Withstand temperatures to 85 °C (185 °F)
These ties are injection-molded of 6/6 nylon, and won’t corrode or stretch. Durable locking mechanism holds securely even when used to bundle vibrating or flexing materials. Ties continue to hold tension even in oily spray or wet environments. Easily apply ties by hand or use a tensioning tool (tools must be ordered separately).
Ties are UL-component recognized, and are self-extinguishing as per UL 94 flam. Class: V-2.
Ties withstand temperatures to 85 °C (185 °F) and have a melting point of 257 °C (495 °F). All ties remain flexible to −40 °C (−104 °F). Efficient range of ties may be higher or lower depending upon specific applications.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty