Would you like to log in again?
VWR® Plus Series Compact Laboratory Refrigerators with Natural Refrigerants
Supplier: VWR
VWR®
VWR-HC-10PS VWR-HC-10PG
76580-148EA 4226.76 USD InStock
76580-148 76580-150
VWR® Plus Series Compact Laboratory Refrigerators with Natural Refrigerants
Refrigerators
These VWR® Plus Series compact laboratory refrigerators offer premium cold storage with superior temperature control.
  • Microprocessor temperature controller
  • Digital temperature display
  • Audible and visual alarm systems
  • LED interior lighting, and adjustable shelving
  • Keyed door lock
  • Natural, hydrocarbon refrigerant
  • ENERGY STAR certified*
These compact glass door laboratory refrigerators offer professional cold storage for scientific and industrial settings. Units come equipped with temperature alarms, keyed door locks, forced draft circulation, probe access, and precise temperature monitoring through microprocessor control. The Plus Series line includes premium additions such as extensive alarm systems and digital touch pad displays. All units run on natural, hydrocarbon refrigerant for environmental health and energy efficiency.
Material: White powder coat
Certifications: UL/C-UL listed.
*Select models ENERGY STAR certified: 76580-148, 76580-150
Ordering information: All units come with a two year parts and labor warranty, plus an additional three year compressor parts warranty.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty