Would you like to log in again?
VWR® Standard Laboratory Freezers Solid Door Manual Defrost with Natural Refrigerants
Supplier: VWR
VWR®
VWR-HC-MFP-20 VWR-HC-MFP-17 VWR-HC-MFP-14
76580-174EA 4376.01 USD InStock
76580-174 76580-172 76580-170
VWR® Standard Laboratory Freezers Solid Door Manual Defrost with Natural Refrigerants
Freezers Laboratory / General Purpose Freezers
These VWR® Standard Series manual defrost freezers offer laboratory cold storage at an economical pricepoint.
  • Microprocessor temperature controller
  • Digital temperature display
  • Audible and visual alarm systems
  • Interior shelving
  • Keyed door lock
  • Natural, hydrocarbon refrigerant
These manual defrost laboratory freezers offer professional cold storage for scientific and industrial settings. Units come equipped with temperature alarms, keyed door locks, forced draft circulation, probe access, and precise temperature monitoring through microprocessor control. The Standard line provides multi-functional features in a cost-effective design. All units run on natural, hydrocarbon refrigerant for environmental health and energy efficiency.
Material: White powder coat
Certifications: UL/C-UL or ETL Listed.
Ordering information: All units come with a two year parts and labor warranty, plus an additional three year compressor parts warranty.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty