Would you like to log in again?
VWR® Plus Laboratory Freezers with Manual Defrost (30 cu. ft.)
Supplier: Avantor
VWR®
VWR-MFP-3030 VWR-MFP-30
76580-196EA 14407.6 USD InStock
76580-196 76580-194
VWR® Plus Laboratory Freezers with Manual Defrost (30 cu. ft.)
Freezers Laboratory / General Purpose Freezers
These VWR® Plus Series manual defrost freezers offer premium cold storage with superior temperature control.
  • Microprocessor temperature controller
  • Five inner "fast-freeze compartments" with doors
  • Digital temperature display and alarm module
  • Audible, visual, senor error, and power failure alarms
  • Alarm validation and alarm mute with ring back
  • Keyed door lock
These manual defrost laboratory freezers offer professional cold storage for scientific and industrial settings. Units come equipped with temperature alarms, keyed door locks, forced draft circulation, probe access, and precise temperature monitoring through microprocessor control. The Plus Series line includes premium additions such as extensive alarm systems and digital touch pad displays. All units run on natural, hydrocarbon refrigerant for environmental health and energy efficiency. A ‒30 °C option is available.
Material: White powder coat
Certifications: UL/C-UL listed.
Ordering information: All units come with a two year parts and labor warranty, plus an additional three year compressor parts warranty.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty