Would you like to log in again?
Haug Anti-Static Brush, Mettler-Toledo
Supplier: Mettler-Toledo
63052301
76580-228EA 31.25 USD InStock
76580-228
Haug Anti-Static Brush, Mettler-Toledo
Brushes
Anti-static brush to remove electrostatic charges, prevent cross contanimation, and enable accurate weighing results.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty