Would you like to log in again?
FitMe™ Lab Coats
Supplier: Dukal
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
FitMe™
UGC-6603-XXL UGC-6609-XXL UGC-6602-M UGC-6602-XL UGC-6605-M UGC-6613-XXL UGC-6603-S UGC-6602-S UGC-6600-XL UGC-6605-XXL UGC-6602-L UGC-6600-S UGC-6609-XL UGC-6603-L UGC-6605-XL UGC-6613-M UGC-6603-XL UGC-6613-L UGC-6601-S UGC-6604-XXL UGC-6602-XXL UGC-6610-XL UGC-6610-XXL UGC-6601-XXL UGC-6601-M UGC-6604-M UGC-6613-XL UGC-6603-M UGC-6610-S UGC-6600-XXL UGC-6600-M
76582-160CS 209.41 USD InStock
76582-160 76582-182 76582-282 76582-180 76582-186 76582-242 76582-220 76582-162 76582-262 76582-168 76582-268 76582-224 76582-188 76582-166 76582-200 76582-228 76582-248 76582-208 76582-190 76582-192 76582-198 76582-272 76582-196 76582-278 76582-256 76582-114 76582-210 76582-232 76582-156 76582-134 76582-112
FitMe™ Lab Coats
Workwear
FitMe™ lab coats feature a unique adjustable waistband design that provides a true custom fit.
  • Knit cuffs and collar
  • Snap front closure
  • Premium SMS material
  • 2 hip pockets, 1 chest pocket
  • Washable and autoclavable up to 5 times
Available in a wide variety of colors, FitMe™ lab coats are manufactured from premium SMS material, and are supplied non sterile.
Packaging: Lab coats are packaged 10 per bag, 5 bags per case.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty