Would you like to log in again?
FitMe™ Lab Jackets
Supplier: Dukal
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
FitMe™
UGJ-6504-XL UGJ-6509-L UGJ-6500-XXL UGJ-6505-L UGJ-6500-L UGJ-6504-XXL UGJ-6500-S UGJ-6501-XL UGJ-6509-M UGJ-6504-S UGJ-6503-S UGJ-6509-XL UGJ-6502-XL UGJ-6505-M UGJ-6500-M UGJ-6504-L UGJ-6502-XXL UGJ-6505-S UGJ-6501-L UGJ-6501-M UGJ-6503-M UGJ-6509-XXL UGJ-6510-XXL UGJ-6509-S UGJ-6513-L UGJ-6503-L UGJ-6501-S UGJ-6513-M UGJ-6500-XL UGJ-6505-XL UGJ-6510-S UGJ-6504-M UGJ-6503-XXL UGJ-6502-S UGJ-6503-XL UGJ-6513-XL UGJ-6501-XXL UGJ-6510-XL UGJ-6510-M UGJ-6502-M UGJ-6510-L UGJ-6513-S UGJ-6502-L UGJ-6513-XXL UGJ-6505-XXL
76582-280CS 198.28 USD InStock
76582-280 76582-164 76582-286 76582-264 76582-184 76582-284 76582-240 76582-146 76582-246 76582-122 76582-144 76582-244 76582-222 76582-128 76582-148 76582-126 76582-226 76582-108 76582-270 76582-150 76582-172 76582-194 76582-170 76582-252 76582-230 76582-274 76582-132 76582-176 76582-110 76582-130 76582-152 76582-174 76582-212 76582-234 76582-158 76582-254 76582-276 76582-178 76582-238 76582-216 76582-118 76582-214 76582-236 76582-116 76582-218
FitMe™ Lab Jackets
Workwear
FitMe™ lab jackets feature a unique adjustable waistband design that provides a true custom fit.
  • Knit cuffs and collar
  • Snap front closure
  • Premium SMS material
  • 2 hip pockets, 1 chest pocket
  • Washable and autoclavable up to 5 times
Available in a wide variety of colors, FitMe™ lab jackets are manufactured from premium SMS material, and are supplied non sterile.
Packaging: Lab jackets are packaged 10 per bag, 5 bags per case.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty