Would you like to log in again?
Cole-Parmer® BM-600 Planetary Mill High-Energy Ball Mill
Supplier: SPEX SamplePrep
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Cole-Parmer®
8227B-10 8245 8221B-40 8221 8221B-30 8211S 8221B-10 8200-115 8240 8221B-20 8211 8227 8227B-40 8227B-20 8217 8221S 8227B-30
76582-292EA 2359.41 USD InStock
76582-292 76582-290 76582-296 76582-294 76582-300 76582-312 76582-298 76582-320 76582-310 76582-288 76582-304 76582-316 76582-302 76582-314 76582-308 76582-306 76582-318
Cole-Parmer® BM-600 Planetary Mill High-Energy Ball Mill
Mills
The Cole-Parmer® BM-600 planetary mill high-energy ball mill grinds samples with a mechanical motion rotating the jar in one direction while the platform, planetary wheel, often referred to as the sun wheel, is rotated in the opposite direction. The mechanical milling is performed at a 2:1 ratio causing the jar to rotate twice for every single rotation of the sun wheel. As the jar is moved, the relative centrifugal force is transferred to the grinding balls causing them to move in a circular motion, into each other and against the jar wall, grinding the sample. Functionally described as a high-shearing or a high-energy ball mill, this mill is used for pulverizing rocks, minerals, sand, cement, slag, ceramics, glass, and other brittle to hard samples to analytical fineness, blending powders or making emulsions.
The mills also feature a touch screen display, sleek modern design, forced-air cooling, a safety interlock, and a choice of hardened steel or stainless steel grinding jars and media, which are purchased separately.
Certifications: CE approved.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty