Would you like to log in again?
Sol-M® Blunt Fill Needles
Supplier: Sol-Millennium
Sol-M®
110022 110021 110022F
76582-564BX 41.31 USD InStock
76582-564 76582-356 76582-354
Sol-M® Blunt Fill Needles
Syringe Needles
The Sol-M Blunt Fill Needle reduces the risk of needlestick injuries to healthcare workers when drawing up and preparing medication by removing the need for sharps.
  • Eliminates the unnecessary use of sharps
  • Color-coded hubs for easy identification
  • Needle hub is compatible with all Luer Lock and Luer Slip Tip syringes
  • Available with 5-micron filter to prevent the drawing of glass particles when using ampules
This can be used with the ‘Double Needle Technique’ when drawing medication and can also be used for IV push administration in needleless IV systems.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty