Would you like to log in again?
Sol-Care® Safety Winged Blood Collection Needles, Sol M
Supplier: Sol-Millennium
Sol-Care®
110201030033 110201030031 110201030002 110201030034 110201030030 110201030012 110201030006 110201030032 110201030004 110201030010 110201030035 110201030008
76582-472BX 92.05 USD InStock
76582-472 76582-462 76582-470 76582-460 76582-454 76582-476 76582-466 76582-474 76582-464 76582-458 76582-456 76582-468
Sol-Care® Safety Winged Blood Collection Needles, Sol M
Syringe Needles
This safety-engineered device has a sliding safety shield that clicks when activated, alerting you that the needle is safely locked.
  • Transparent design that allows visualization of the blood flashback and flow
  • Single-handed activation option that provides more flexibility to care for the patient and venipuncture site
  • Sliding safety shield pushes over the needle and locks with an audible click, protecting against needlestick injury
Available with or without pre-attached holder.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty