Would you like to log in again?
Sol-Vet® Blood Collection Needles (Veterinary Care), Sol M
Supplier: Sol-Millennium
Sol-Vet®
110301070003 110301070001 110301070004 110301070002
76582-502BX 10.28 USD InStock
76582-502 76582-500 76582-506 76582-504
Sol-Vet® Blood Collection Needles (Veterinary Care), Sol M
Syringe Needles
The Sol-Vet Butterfly Needle features a removeable multi-sample luer adapter, making its application flexible across multiple uses. This is a safety-engineered device with a sliding safety shield that clicks when activated, telling you the needle is safely locked.
  • Thin-walled needle minimizes patient discomfort
  • Transparent design allows for clear visualization of blood flow
  • Available with or without pre-attached blood collection tube holder
Products for veterinary use only.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty