Would you like to log in again?
Sol-Care® Safety Insulin Syringes, Sol M
Supplier: Sol-Millennium
100089IM 100017IM 100071IM 100081IM 100002IM
76582-508BX 43.86 USD InStock
76582-508 76582-532 76582-514 76582-556 76582-548
Sol-Care® Safety Insulin Syringes, Sol M
Syringes Syringes, Medical
This Insulin syringe provides accurate and comfortable insulin administration, with features to help prevent needlestick injuries.
  • Needles are tri-bevel and equipped with plunger lock and break feature that prevents reuse of syringe
  • Low dead space - improved accuracy and less medication waste when compared to conventional syringes
  • Bold and clear graduated insulin unit markings for accurate dosing
Safety feature: Needle retracts into the syringe barrel as a safety feature to prevent needlestick injuries.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty