Would you like to log in again?
Sol M® Oral Syringes (Gasket Type, Bulk NS)
Supplier: Sol-Millennium
21010GCNSB 21005GCNSB 21003GANSB 21003GCNSB 21001GCNSB 21010GANSB 21001GANSB 21005GANSB
76582-682CS 92.94 USD InStock
76582-682 76582-706 76582-690 76582-680 76582-696 76582-698 76582-688 76582-704
Sol M® Oral Syringes (Gasket Type, Bulk NS)
Syringes Syringes, Medical
The Sol-M oral syringe features a gasket design, and has been specifically designed for use with oral medications.
  • Clear graduation markings for accurate medication delivery
  • Not made with natural rubber latex
  • Optional amber barrel designed for light-sensitive drugs, in compliance with United States Pharmacopeia (USP) requirements
Special tip design will not accept Luer connectors, preventing medication errors and the accidental intravenous injections of oral liquids.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty