Would you like to log in again?
Sol-M® Syringe Convenience Trays
Supplier: Sol-Millennium
110101120011 1832110T 1832210T P180003TU P180005TU P180030TU P180010TU P180011T P180020TU 180001T 1852110T 1832310T P180060TU 110101120012 1852010T
76582-816CS 97.97 USD InStock
76582-816 76582-652 76582-650 76582-810 76582-436 76582-656 76582-434 76582-830 76582-798 76582-804 76582-826 76582-628 76582-823 76582-658 76582-648
Sol-M® Syringe Convenience Trays
Syringes Syringes, Medical
The convenience trays are intended to be used in pharmacies to aid with batch-filling.
  • Clear, bold graduations for accurate filling and dosing
  • Sterile trays for convenient filling and use
  • Available with safety-engineered syringes
A wide variety of Sol-Millennium's syringes are available in sterile convenience trays. Intended to be used in pharmacies, multiple syringes are packaged in sturdy trays that are ready to fill and use. Both standard and safety-engineered syringes are available in convenience trays.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty