Would you like to log in again?
Sol-Care® Safety Multi-Sample Needles, Sol M
Supplier: Sol-Millennium
Sol-Care®
SMSNH21125 SMSNH22125 SMSN22125 SMSN21125
76582-906BX 33.22 USD InStock
76582-906 76582-904 76582-902 76582-900
Sol-Care® Safety Multi-Sample Needles, Sol M
Syringe Needles
The Sol-Care Safety Multi-Sample Blood Collection Needles are designed with a safety shield that completely covers and locks over the needle after use to protect against needlestick injuries.
  • Safety shield aligns with a bevel up needle orientation, allowing for low-angle penetration
  • Pivoting safety shield with audible click that indicates it is locked in place
  • Safety mechanism is designed for one-handed activation
  • Safety shield is color coded to reduce potential errors
Compatible with all standard holders.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty