Would you like to log in again?
Eppendorf Tubes® Microtubes with Screw Cap, 5.0 ml
Supplier: Eppendorf
Eppendorf Tubes®
0030122313 0030122321 0030122518 0030122518 0030122330 0030113500 0030122402 0030113500 0030122305
76587-088PK 106.7 USD InStock
76587-088 CA76587-088 CA77391-926 77391-926 75874-640 76019-018 75874-636 76313-962 75874-638
Eppendorf Tubes® Microtubes with Screw Cap, 5.0 ml
Tubes Microcentrifuge Tubes
Eppendorf Tubes® with screw cap are an ideal choice when working with medium-sized sample volumes (0.5 to 5.0 ml). SafeCode variants enhance sample identification, while the BioBased option, manufactured using renewable resources, provides an eco-friendly alternative without compromising performance.
  • Polypropylene (PP) tubes, high-density polyethylene (HDPE) screw caps
  • Ensures leak-free storage and protects sample integrity
  • Suited for a wide range of laboratory tasks, enhancing flexibility
  • High-quality materials for long-lasting, robust performance
  • Clear, visible graduations for quick volume assessment
  • Facilitates optimal sample recovery and efficient centrifugation
  • Available in Eppendorf quality, PCR clean, Sterile/Pyrogen-/DNase-/RNase-/Human and Bacterial DNA-free, and Forensic DNA grade quality for tested purity
BioBased Option:
The Eppendorf Tubes® BioBased options open up new doors to make laboratory work significantly more sustainable without jeopardizing experimental results. By using a second-generation biobased polymer made from 90% renewable resources (e.g. waste and residues from vegetable oil refining or waste cooking oil) the product-related carbon footprint of these tubes is significantly reduced. BioBased varients aid in the reduction of consumable-related carbon footprint in the lab. The bio-based tubes have identical material properties and technical specifications as non-BioBased tubes.
2D SafeCode Options (Barcoded):
A clean and stable label on the side or on the top of the vessel can make scientific life so much easier. Eppendorf tubes are available with SafeCode, a well-designed 2D barcoding system. Take advantage of the pre-labeled and durable 2-level coding of the SafeCode system (datamatrix code and clear coding) to enable safe sample identification. The safe sample ID is supported by additional online information about the dedicated consumable at the Eppendorf dataport.
A barcode is an optical representation of data which is machine-readable. Classic 1D barcodes code data by varying the width of the parallel lines as well as their spacings. Two-dimensional codes (2D) are based on dots, hexagons, rectangles, or other geometric patterns. These codes are also called data matrix codes. ECC200 error correction function enables safe code reading with up to 25% damaged codes.
Manufactured without the use of slip agents, plasticizers, and biocides – substances that have been shown to leach from plastic consumables into the sample and negatively affect bio-assay results.
Certifications: ACT label, Lot-specific certificates for the purity grade PCR clean, Sterile, Forensic DNA Grade; ISCC PLUS declarations (BioBased Varient) can be downloaded from the Eppendorf website.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty