Would you like to log in again?
3M™ Harvest RC Single-Stage Chromatographic Purification Systems
Supplier: 3M Healthcare
3M™
7100236845 7100236865 7100241969 7100236866 7100236867 7100245350 7100242700 7100236863
76587-110CS 1882.4 USD InStock
76587-110 76587-124 76587-104 76587-126 76587-114 76587-128 76587-116 76587-118
3M™ Harvest RC Single-Stage Chromatographic Purification Systems
Protein Purification Protein Purification Equipment
Single-stage chromatographic purification for recombinant protein therapeutic manufacturing.
  • High mAb product recovery (capsules: >95%; conical tube and well plate >90%)
  • Consistent cell loading capacity
  • Turbidity reduction (<15 NTU)
  • TDNA reduction (<500 ppb)
  • Minimal cell shear
  • 0.1 μm sterile filter protection
The first step in the recombinant biotherapeutic process is harvesting cell culture fluid containing the product. Conventional approaches for performing this unit operation utilize a combination of depth filtration, centrifugation and membrane filtration. These technologies utilize differences in density and in size to yield higher cell densities and product titers, the ability to effectively harvest the cell culture fluid with the consistency and scalability required becomes challenging.
3M Harvest RC is a harvest solution that utilizes fibrous anion exchange (AEX) chromatography to efficiently separate the cells, cell debris and DNA from the harvest fluid containing the target product. Precision quaternary ammonium (Q) functionalized polypropylene fiber, combined with a 0.2 µm PES membrane, provides scalable and predictable clarification from discovery to commercial manufacturing scale.
3M™ Harvest RC chromatographic clarifier encapsulates innovative synthetic fibrous anion exchange (AEX) chromatography media and a 0.2 µm polyether sulfone (PES) membrane. This enables a single-stage clarification process of low to high-density cell culture (>40 million cells per ml) with high product recovery and high fidelity of soluble and insoluble contaminant separation.
Cells are bound inside the media by electrostatic charge interaction with the AEX chromatographic fibers. This results in the efficient retention of large and small particulates without developing a surface cake layer. The media can also remove soluble impurities which results in cleaner effluent than centrifugation or depth filtration.
Certifications: Meet cGMP requirements (for BC340, BC1020, BC2300 and BC16000) or GLP requirements (for CT15, WP6, BC4 and BC25).
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty