Would you like to log in again?
Gamma Irradiated Sterility Maintenance Covers, Heat Seal
Supplier: Keystone Cleanroom Products
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
3235ST 3220ST 3222ST 3230ST 3215ST 3234ST
76595-182CS 2423.28 USD InStock
76595-182 76595-196 76595-198 76595-186 76595-178 76595-206
Gamma Irradiated Sterility Maintenance Covers, Heat Seal
Bags Sterilization Pouches
Ideal for pharmaceutical and biotech production applications.
  • Wide range of sizes
  • Works well with impulse and rotary heat sealers
  • No tools required to open
  • Impervious to liquid sanitization agents
  • Visibility inside the bag allows for identification
  • Prolong the storage and shelf-life of products sterilized 'in-house'
After unloading the autoclave, under Grade A LAF, place wrapped, steam sterilized items into sterility maintenance bags and seal closed. This technique captures the conditions of Grade A and protects sterilized products from external sources of contamination during storage and transport through lesser classified areas.
Certifications: Certificates of Irradiation and Certificates of Conformance sent with shipments.
Packaging: The sterility maintenance covers are packaged in Tyvek Chevron peel pouches with Lot # and expiration date for traceability.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty