Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Gamma Irradiated Sterility Maintenance Covers, Self-Seal, Keystone Cleanroom Products
Supplier: Keystone Cleanroom Products
3320ST 3320ST3 3310ST 3334ST 3335ST 3315ST 3334ST3 3330ST 3322ST3 3322ST 3312ST 3315ST5
76635-472CS 1319.18 USD InStock
76635-472 76635-470 76595-184 76595-176 76595-200 76635-474 76595-188 76595-202 76595-204 76595-190 76595-192 76595-180
Gamma Irradiated Sterility Maintenance Covers, Self-Seal, Keystone Cleanroom Products
Bags Sterilization Pouches
Ideal for pharmaceutical and biotech production applications.
  • Wide range of sizes
  • Convenient self-seal closure; no need for the use of or validation of a heat sealer
  • No tools required to open
  • Impervious to liquid sanitization agents
  • Visibility inside the bag allows for identification
  • Prolong the storage and shelf-life of products sterilized ‘in-house’
After unloading the autoclave, under Grade A LAF, place wrapped, steam sterilized items into sterility maintenance bags and seal closed. This technique captures the conditions of Grade A and protects sterilized products from external sources of contamination during storage and transport through lesser classified areas.
Certifications: Certificates of Irradiation and Certificates of Conformance sent with shipments.
Packaging: The sterility maintenance covers are packaged in Tyvek Chevron peel pouches with lot number and expiration date for traceability.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty