Would you like to log in again?
Flextril™ Nitrile Chemical Gloves
Supplier: Honeywell Safety
33-3150E/9L 32-3011E/7S/N 32-3011E/11XXL/N 33-3150E/10XL 32-3015E/7S 33-3765E/11XXL 33-3765E/7S 33-3765E/9L 32-3015E/9L 32-3015E/8M 32-3015E/10XL 33-3150E/8M 33-3150E/11XXL 33-3150E/7S 33-3765E/10XL 33-3765E/8M 32-3011E/8M/N
76595-240BG 166.88 USD InStock
76595-240 76595-242 76595-220 76595-230 76595-244 76595-222 76595-232 76595-224 76595-246 76595-234 76595-226 76595-214 76595-236 76595-228 76595-238 76595-216 76595-218
Flextril™ Nitrile Chemical Gloves
Gloves
Flextril™ gloves are a hand protection solution designed to provide fivefold performance – high chemical protection, reliable cut resistance, great wet and dry grip, enhanced dexterity, and improved comfort. Additionally, all glove models feature a printed QR code, providing all of the necessary performance data at the wearer's fingertips.
  • Superior chemical and cut protection
  • Great wet and dry grip
  • Enhanced dexterity and comfort
  • Sweat management features
  • Cost-effective
  • Increased durability
  • Food contact compliant
The Flextril™ nitrile barrier protects hands against chemicals, irritants, or other potentially hazardous materials, while the cut-resistant technology shields against lacerations. The specially designed coating gives users the grip performance that is needed when wearing chemical gloves. Handle tools even more efficiently with an optimal thickness that offers enhanced dexterity and increased ease of movement. Moreover, the ergonomic design delivers high-performance hand support and comfort.
Flextril™ gloves feature an innovative QR code printed on each glove, providing access to all of the technical specifications associated with each model. This includes chemical permeation data, coating and liner materials, thickness, technical certifications and sizes. Simply scan the QR code printed on each glove to determine the exact model for virtually any application.
Flextril™ gloves may be used in a wide variety of applications, including use in the automotive industry, chemical manufacturing, machinery and equipment, oil and gas, metal fabrication, and food processing.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty