Would you like to log in again?
Induro Reverse Transcriptase
Supplier: New England Biolabs
M0681S M0681L M0681S M0681L
76595-286EA 198 USD InStock
76595-286 CA76595-274 CA76595-286 76595-274
Induro Reverse Transcriptase
Enzymes
Induro reverse transcriptase is a group II intron-encoded RT that exhibits increased thermostability, high processivity, and increased tolerance of inhibitors in the synthesis of cDNA from RNA.
  • Highly processive enables fast reactions (>12 kb in under 10 min)
  • Supports direct RNA sequencing and long read cDNA sequencing workflows
  • Excellent tolerance of common inhibitors in RNA biological samples
  • Increased thermostability for robust cDNA synthesis at high temperatures (55 °C)
  • No DTT requirement for reaction setup
It is an ideal enzyme for challenging cDNA synthesis reactions from long transcripts, RNAs with strong secondary structures, and RNA samples with inhibitors. Additionally, induro reverse transcriptase supports RNA-seq applications such as long-read RNA sequencing.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty