Would you like to log in again?
Msz Exonuclease I
Supplier: New England Biolabs
M0527S M0527S
76595-278EA 78 USD InStock
CA76595-278 76595-278
Msz Exonuclease I
Enzymes
A DNA specific exonuclease isolated from Methylocaldum szegediense that catalyzes the removal of nucleotides from linear single-stranded DNA in the 3' to 5' direction with optimal activity between 45 and 60 °C. Isolated from a strain of E. coli which carries the cloned pMC45-1 gene from Methylocaldum szegediense.
  • Catalyzes removal of nucleotides from linear single-stranded DNA in 3' to 5' direction
  • Ideal for thermophilic workflows between 45 and 60 °C
  • Can be heat inactivated at 80 °C in 1 minute
  • Supplied with 1 vial of rCutSmart Buffer (10X)
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty