Would you like to log in again?
Q5 Blood Direct 2X Master Mix, New England Biolabs
Supplier: New England Biolabs
M0500S M0500L M0500S M0500L
76595-272EA 131 USD InStock
76595-272 CA76595-280 CA76595-272 76595-280
Q5 Blood Direct 2X Master Mix, New England Biolabs
Nucleic Acid Reagents End-point PCR Enzymes and Kits
Q5 Blood direct 2X master mix can amplify a wide variety of targets direct from dried blood spots to up to 30% whole human blood, skipping DNA purification. The master mix includes polymerase and dNTPs in an optimized buffer with increased resistance to inhibitors in blood, anti-coagulants, and chemicals on filter papers. It is capable of amplifying products up to 7.5 kb from human whole blood cells preserved with Na EDTA, K2 EDTA, Na citrate and Na heparin as well samples stored on common preservative filter papers.
  • Amplify direct from whole blood cells or dried blood spots without DNA extraction or purification
  • Features Q5 hot start high-fidelity DNA polymerase for accurate and efficient PCR
  • Optimized buffer for increased resistance to inhibitors and chemicals
  • Consistent performance with a variety of filter types and anti-coagulants present
  • Aptamer-based enzyme control enables room temperature reaction setup
The master mix features Q5 hot start high-fidelity DNA Polymerase, a thermostable DNA polymerase with 3' to 5' exonuclease activity, fused to a processivity-enhancing Sso7d domain for robust DNA amplification with low error rates. The addition of an aptamer-based inhibitor allows for room temperature reaction setup.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty