Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Sartolab® Storage Bottles, Single-Use, Polystyrene, Sartorius
Supplier: Sartorius
Sartolab®
18023E 18022E 18025E 18024E
76595-310PK 72.2 USD InStock
76595-310 76595-312 76595-326 76595-306
Sartolab® Storage Bottles, Single-Use, Polystyrene, Sartorius
Bottles Plastic Bottles
Single-use polystyrene bottles for storage of media, buffers and other aqueous solutions.
  • Low-profile design for easy gripping with one hand
  • Ergonomically designed cap to provide a rigid grip and one-hand handling of each bottle
  • The bottles with an indented bottom are stackable to save space in the refrigerator and in the freezer
  • Caps (45 mm) with airtight seal to minimize the risk of contamination
  • Graduation marks on the bottles ensure accuracy and easy reading
  • Designated writing field for clear labeling of samples
  • The materials used in the production are medical-grade for the highest quality, without any animal origin
  • Sterile and endotoxin-free
Sartorius single-use polystyrene (PS) storage bottles are designed for safe, secure storage of tissue culture media and sera, buffers and other aqueous solutions. The bottles can be used with all Sartolab® RF / BT vacuum filtration units with a 45 mm neck size.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty