Would you like to log in again?
Oakton® ORPTestr® 30 Double Junction Waterproof Pocket Tester, Antylia Scientific
Supplier: Antylia Scientific
Oakton®
35634-32
76595-382EA 108.24 USD InStock
76595-382
Oakton® ORPTestr® 30 Double Junction Waterproof Pocket Tester, Antylia Scientific
Electrochemical Meters pH and Multi-Parameter Meters
Save time and money with replaceable double-junction sensor.
  • Easy-to-navigate interface for quick setup and calibration
  • Redesigned leakproof cap functions as a sample, calibration, and hydrating cup
  • Auto-shutoff function conserves battery life
  • IP67-rated housing is waterproof and floats when sensor cap is on
The Oakton® ORP 30 Tester features a replaceable double-junction platinum electrode ideal for checking dirty water, heavy metals, sulfides, and organics. Plastic electrode body resists most chemicals.
Applications range from swimming pool testing to pulp bleaching. Meter features ±2 mV accuracy and a ±150 mV offset. Housing allows you to attach a lanyard (included) for easy retrieval. Features include large easy-to-read LCD with 3-color backlight, a Hold function, stable reading indicator, self-diagnostic error messages, and a three-segment battery-life indicator. Leakproof cap provides stabilizing base to stand the meter upright.
Delivery information: Protective plastic storage case, lanyard and batteries.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty