Would you like to log in again?
Moticam® X5 Digital Camera Wi-Fi, Motic
Supplier: Motic
Moticam® Motic Moticam
1100600101322 1100600101322
76595-460EA 436.77 USD InStock
76595-460 CA76595-460
Moticam® X5 Digital Camera Wi-Fi, Motic
Cameras
The Moticam® X5 is a high-resolution streaming Wi-Fi camera. By creating its own Wi-Fi signal, simply connect with your android or iOS device and view, capture, and edit live images from your microscope with our free MotiConnect app.
  • Attachable digital microscopy camera with built-in Wi-Fi signal
  • Transmits to Wi-Fi tablets and Wi-Fi computers
  • 2 MP streaming live resolution over Wi-Fi
  • 4 MP CMOS sensor and captured resolution
  • Motic Images Plus 3.1 for Windows/macOS
A powerful high-resolution to turn your microscope into a Wi-Fi station. Simply logon with your tablet or smartphone and get ready to capture, edit and save your images with the free MotiConnect app. Use your computer with motic image plus 3.1 Software for further image processing.
Delivery information: CS-ring adaptor, focusable Lens, 30 and 38 mm eyepiece adapters, USB 2.0 cable, Motic 4-Dot calibration slide, macro tube, calibration card included.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty