Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Moticam® BTI8 Tablet Digital Camera, Motic

Supplier: Motic
Moticam® Motic Moticam
1100600101501 1100600101501
76595-462EA 776.25 USD InStock
76595-462 CA76595-462
Moticam® BTI8 Tablet Digital Camera, Motic
Cameras
The Moticam BTI8 combines a space-saving camera hardware with new Wi-Fi technology. Use the pre-installed MotiConnect App and get ready to capture, edit and share your microscope images from the tablet in a few taps.

  • 8" LCD android tablet with 1280×800 screen resolution
  • Connection: Wi-Fi (2.4 and 5 GHz), ethernet
  • Wi-Fi resolution: 4.0 MP captured video resolution
  • Wi-Fi: Wi-Fi output (802.1 a/b/g/n/ac supporting both hot-spot mode or using existing Wi-Fi network)
  • Preloaded optimized MotiConnect app

Moticam® BTI is the most flexible tablet solution, consisting of a 4 MP camera with a built-in adjustable bracket that allows you to mount your own tablet. The camera can be connected via Wi-Fi to an iOS or Android tablet. The Moticam BTI will help microscope users visualize, edit and share the microscope images, enabling computer-free digital microscopy.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty